Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WAY 170523, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
Description
Potent and selective inhibitor of MMP-13 (IC50 values are 17, 945, > 1000 and > 10000 nM for MMP-13, MMP-9, TACE and MMP-1 respectively).
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 613.68 |
Percent Purity | >99% |
Chemical Name or Material | WAY 170523 |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction