Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WEB 2086, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
Description
Potent, selective platelet-activating factor (PAF) receptor antagonist (Ki = 16.3 nM). Displays anti-inflammatory, antiangiogenic and anticancer activity. Inhibits growth and proliferation of MCF-7 breast cancer cells.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 455.96 |
Percent Purity | >99% |
Chemical Name or Material | WEB 2086 |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction