Learn More
Terumo BCT Inc SCD IIB Weld Integrity Test Kit
Supplier:  Terumo BCT Inc 62002QTY 3+
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Simplify the process of satisfying quality assurance and Good Manufacturing Practices (GMP) requirements. Perform test welds on-site without shipping your welder. Return your test welds to us to evaluate alignment, leakage, and tensile strength
Catalog No. NC1976747
missing translation for 'QuantityLeft'