Learn More
Wells Lamont™ MT130 Sterile/Non-Sterile Touchscreen Cut Resistant Gloves
Catalog No. p-4961183 Shop All Wells Lamont Industry Group Products
Click to view available options
Glove Cuff Color:
This item is not returnable. View return policy
This item is not returnable. View return policy
Ultra-lightweight construction provides bare hand dexterity and superior tactile sensitivity
  • Patent pending yarn technology delivers cut resistance and touchscreen capability
  • Minimal particle shedding to meet cleanroom classifications and requirements
  • Meets Category II using Helmke Drum Test referenced in IEST –RP-CC003.3
  • ANSI Level 4
  • Ambidextrous gloves available in different sizes
Specifications
Color
Gray
Product Type
Cut-resistant Gloves
Size
X-Small
Glove Cuff Color
Gold