Learn More
Electron Microscopy Sciences Blu-Lite UHC™ Western Blotting Film, 5 x 7"
Supplier:  Electron Microscopy Sciences 7047301
Encompass_Preferred
Blu-Lite UHC high contrast, high sensitivity autoradiography film is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications.
Blu-Lite UHC's is also coated on both sides (double emulsion) for improved consistency and overall performance. Especially useful with chemiluminescence, BluBlot™ HS works well with 14C, 32P, 125I, and 35S. This new film reliably produces sharp images which allow for maximum DNA visualization with unsurpassed clarity.
Compatible with automatic processors and manual development methods.
Size: 5 x 7"
Catalog No. 50-193-1059
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.