Learn More
Teknova WFI Quality Water, 200L bag
Supplier: Teknova 20143
WFI Quality Water is a high-grade water that meets both EP and USP grade specifications and is 0.1 µm sterile filtered at the time of fill. It is ideal for producing cell culture media, reconstitution of biochemical reagents, or as a final-rinse water for critical applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.