Learn More
Biotium Wheat Germ Agglutinin, CF350 Conjugate
Supplier: Biotium 290211
This product is fluorescently-labeled wheat germ agglutinin (WGA). WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine. WGA conjugates can be used to stain glycoproteins that have these sugar moieties. WGA can be used as a gram stain to fluorescently label gram bacteria but not gram- bacteria. WGA also binds to the bud scars on budding yeast such as Saccharomyces cerevisiae. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. UV-excitable blue fluorescent CF350 dye has excitation/emission at 347/448 nm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.