Learn More
Bioworld Triticum vulgaris (Wheat) Lectin (WGA) - MagneZoomâ„¢ Kit
Supplier:  Bioworld 201630191
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Technical Specifications:Ligand Source:Triticum vulgarisMatrix: MagneZOOM™ (Paramagnetic Silica Coated Beads)Particle size range: 2-5 µmMatrix attachment: AmideBinding Capacity: 1-2 mg/mLInhibiting/Eluting Sugar:Chitin hydrolysateor GlcNAc with acidor saltCarbohydrate for Elution: 0.2MChitin hydrolysateor GlcNAc with acidor saltBlood Group Specificity: A, B, OForm: 25 mg/mL , PBS (1x) pH 7.4 w/ 0.01% ThimerosalpH stability: 4 – 9Storage: 2 – 8 °C, DO NOT FREEZEApplication note: Calcium is required for binding.Kit contents:• MagneZoom™-WGA, 25mg/ml suspension: 2ml• Binding/Wash Buffer • Elution Buffer• Storage BufferThis Kit Requires MagneZoom™ paramagnetic bead station (20160000-1) for magnetic bead separation.
Catalog No. 50-197-622