Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Peristaltic Pump Tubing: Microcassette Tubing

Catalog No. p-4882435 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Inner:
Diameter (English) Inner:

For use with Wheaton Omnispense™ Elite and Unispense™ PRO peristaltic pumps

For small volume applications

Specifications

Product Type

Microcassette Tubing

Autoclavable

Autoclavable

Diameter (English) Inner

0.004 in.

Diameter (Metric) Inner

0.104 mm

Material

Platinum-cured Silicone

Overall Length (English)

50 ft.

Overall Length (Metric)

1524 cm

For Use With

Microcassette pump head

For Use With (Application)

Microcassette pump head

Sterility

Sterile

Color

Translucent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.