Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers
Shop All DWK Life Sciences Products
Click to view available options
Neck Finish:
15-425
20-400
22-400
24-400
28-400
33-400
Description
- Poly-Seal liner is manufactured from low density polyethylene
- Phenolic
- Screw cap
- Color: Black
Specifications
Specifications
Color | Black |
Material | Phenolic Resin |
No. per Case | 144 Cs. |
Neck Finish | 15-425 |
Certifications/Compliance | ISO 9001 |
For Use With (Equipment) | Redi-pak™ container |
Closure Liner | Polyseal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction