Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ White PTFE/Silicone Septa
Vial septa are available in a variety of materials
Supplier: MilliporeSigma™ Supelco™ 507784
Description
- Septa are selected based on the chemical properties of the sample and the temperature to which the vial will be subjected during the analysis.
- PTFE/Silicone is for use with most organic solvents.
- PTFE/Silicone is not for use with chlorosilanes.
- It has good resealability and can withstand a temperature of 250°C.
Specifications
8 mm | |
PTFE/Silicone | |
100 | |
20 durometer |
White | |
Temperature limit 250°C | |
0.06 in. | |
2 mL Standard Opening Vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction