Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
White/Tan PTFE/Silicone Septa
May be used in headspace solid-phase microextraction (SPME) (lyophilized aqueous extract)
Supplier: MilliporeSigma Supelco 33213
Description
- A gas tight and inert nature septa, which is used for high temperature applications during pretreatment and sample oxidation.
Specifications
8 mm | |
PTFE/Silicone | |
1000 | |
Screw Top vial, 8-425 thread |
Tan, White | |
Temperature limit 250°C | |
0.06 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction