Learn More
Wiley™ Handbook of Laboratory Health and Safety
Easy-to-implement approach to ensuring workplace safety.
Supplier:  Wiley™ 047102628X
Encompass
Catalog No. 17-805-12
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Describes the components of a laboratory safety program and how each component fits into the chemical hygiene plan outlined by the new OSHA Lab Standard.
  • Use to help develop OSHA-required Chemical Hygiene Plan
  • Assists laboratory managers in ensuring that labs are safe and in compliance with OSHA regulations
  • Provides step-by-step guidelines for developing procedures that comply with the regulations
  • Key topics include laboratory hazards, training and documentation, emergency plans and response, chemical exposure, medical surveillance, personal protective equipment, and waste management
  • R. Scott Stricoff and Douglas B. Walters; 1995; 480 pages
    Specifications
    Specifications
    Book
    Handbook of Laboratory Health and Safety
    Safety and Training Materials
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Promotions
    Promotions
    Product Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.