Learn More
Chemplex Industries WINDOW FRAME 0.24 MIL 2.5IN DI
Supplier:  Chemplex Industries 3015
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Mylar(TM), 6.0 u (0.24 mil), 2.5 Dia., In.
Catalog No. 50-249-352
missing translation for 'QuantityLeft'