Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bulwark™ Women's Royal FR Lab Coat with Knit Cuffs
Shop All Red Kap Industries Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
This flame resistant lab coat is designed for functionality, comfort, and safety with a CAT 1 arc rating and NFPA™ 2112 compliant. It comes equipped with rib knit cuffs to keep you protected during movement. Features concealed and reinforced snap front closures for easy donning and doffing, side vents at hem for ease of walking, and four exterior pockets that provide ample storage.
- Certifications Level 1: NFPA 2112, ASTM F1506
- Certifications Level 2: Category 1 Protection
- Hazard : Flash Fire, Arc Flash
- Number of Pockets: 4
- Primary Closure: Concealed Snap/Gripper
- Wash Care : Industrial Laundry - Light Soil
- Fabric: Body: 4.5 oz. (150 g/m2) Nomex™ IIIA Flame Resistant Plain Weave
- Blend: Body: 93% Nomex™ Aramid / 5% Aramid Fibers / 2% Other Fibers
- Pocket: 4 front patch pockets,Side pass through at each hip
- Features: Rib cuffs for protection during movement
Concealed snap front closure to avoid scratching surfaces
Four front mitered style pockets for ample storage
Pass-through at hips for access to underneath - Product Collection: Nomex
- Protection: ATPV Arc Rating: 4.4
- Country of Origin: Imported, Made in USA Fabric
- Gender: Female
Specifications
Specifications
Product Type | Flame Resistant Lab Coat |
Size | X-Small |
Gender | Women's |
Material | Nomex IIIA |
Color | Royal Blue |
Closure Type | Snaps |
Cuff Style | Knit |
Disposable | Reusable |
Sterility | Non-sterile |
Resistance | Flame |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction