Learn More
Fisherbrand™ Wooden Handle Lab Spatulas Set of 7
Spatulas feature stainless steel construction and Wooden handles. Multiple sizes allow for varied laboratory applications.
Description
Spatulas feature stainless steel construction and Wooden handles. Multiple sizes allow for varied laboratory applications.
- Set of 7 Spatulas: 3 in. (7.6 cm), 4 in. (10.2 cm), 5 in. (12.7 cm), 6 in. (15.2 cm), 8 in. (20.3cm), 10 in. (25.4 cm), 12 in. (30.5 cm)
- Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples
- Economical
- Sturdy and durable
- Stainless steel construction
- Riveted wooden handles
Certifications
ISO 9001-02, GMP, cGMP
Specifications
7 | |
Set of 7 spatulas ranging from 3in-12in | |
Multipe use | |
Various Sizes | |
Stainless Steel |
Autoclavable | |
Spatula | |
3 to 12 in. | |
Wood |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.