Learn More
Workrite™ Workrite™ CP Chemical Splash Protection Lab Coat
Catalog No. p-7115405 Shop All Red Kap Industries Products
This item is not returnable. View return policy
This item is not returnable. View return policy
Workrite™FR Chemical Splash Protection Lab Coat. Milliken ShieldCSR chemical splash protection finish resists penetration and repels inadvertent liquid chemical splash.
  • Featuring Milliken ShieldCSR Chemical Splash protection finish resists penetration and repels inadvertent liquid chemical splash
  • Snap front closure for easy on/off
  • Lay-down collar
  • Splash resistant side pass-throughs for easy inner pocket access
  • Pockets include one welt chest pocket on left chest and two large lower front pockets with flaps
Specifications
Product Type Chemical Resistant Lab Coat
Gender Unisex
Color White
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.