Learn More
Brady™ Workstation Data Automation Software
Shop All Brady Corporation ProductsRequires
Internet connection
- Easy guided user interface makes it possible to set up the system in a few minutes without reading manuals
- Works in a few easy steps: monitors for an incoming CSV file, parses the data into pre-defined fields on the label, and automatically starts printing
- End users never touch or modify the templates, so you can rest assured that the data and labels are printed as intended
- Companion software to Brady LabelMark™ labeling software
- 5 license
- Windows™ 7 Professional; Windows™ 7 Ultimate; Windows™ 7 Enterprise; Windows™ 8.1 Enterprise; Windows™ 8.1 Professional
Specifications
Format
E-Media
Product Type
Software
For Use With (Equipment)
BBP™12 Label Printer, BBP™11 Label Printer, BBP™31 Sign and Label Printer, BBP™33 Printer, BBP™35 Label Maker, BBP™37 Label Maker, BMP™41 Printer, BBP™85 Sign and Label Printer, BMP™51 Label Maker, BMP™53 Label Maker, BMP™71 Label Printer, BRADY 1244 Series, BRADY 1344 Series, BRADY IP™ Series, GlobalMark™ Label Maker, BradyPrinter™ 300PR+, BradyPrinter™ PR Plus Series, BRADY 2461/3481/6441 Series, BRADY MVP Plus Series, BRADY X Plus II Series, Ink Jet Printers, Laser Printers
Memory
200 MB, 1.5 Gigahertz (GHZ) Processor or Faster with 2 GB RAM
Resolution
1024 x 768
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.