Learn More
Brady™ Workstation Scan and Print Portable Kit with BMP61 Printer and Scanner
Shop All Brady Corporation ProductsSpecifications
Color
Blue
Display Type
LCD
For Use With (Equipment)
BBP33 Label Printer
Dimensions (L x W)
12 x 9 x 9.5 in.
Color Capability
Monochrome
Includes
BBP33, Code Reader 2600 Scanner, Scan and Print Suite CD
Memory
4 GB
Print Speed
4 in./sec.
Resolution
300 dpi
Operating System
Windows™ 7 (32- or 64-bit);Windows™ 7 Enterprise;Windows™ 7 Professional;Windows™ 7 SPI;Windows™ 7 Ultimate;Windows™ 8 (32- or 64-bit);Windows™ 8 Professional (32 or 64 bit);Windows™ 8.1 (32- or 64-bit);Windows™ 8.1 Enterprise;Windows™ 8.1 Professional;Windows™ 10
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.