Learn More
Ampco Safety Tools Wrench, Hex Key
AMPCO™Hex Key Wrench performs in high-risk and demanding environments.
$15.64 - $38.25
Specifications
Material | Beryllium Copper |
---|---|
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.; WARNING: Cancer - www.P65Warnings.ca.gov
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-174-751
![]() |
Ampco Safety Tools
WH1/16 |
1/16 in. |
Each for $15.64
|
|
|||||
19-174-752
![]() |
Ampco Safety Tools
WH1/8 |
1/8 in. |
Each for $22.80
|
|
|||||
19-174-765
![]() |
Ampco Safety Tools
WH3/16 |
3/16 in. |
Each for $29.95
|
|
|||||
19-174-766
![]() |
Ampco Safety Tools
WH3/32 |
3/32 in. |
Each for $21.15
|
|
|||||
19-174-772
![]() |
Ampco Safety Tools
WH5/32 |
5/32 in. |
Each for $26.95
|
|
|||||
19-174-773
![]() |
Ampco Safety Tools
WH5/64 |
5/64 in. |
Each for $21.50
|
|
|||||
19-174-776
![]() |
Ampco Safety Tools
WH7/32 |
7/32 in. |
Each for $38.25
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Beryllium Copper | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.