Learn More
Volu Sol Wright Stain, 16 oz / 500 mL, Volu-Sol™
Supplier: Volu Sol VWS016
Romanowsky stain, for peripheral blood and bone marrow smears. Also used to perform differential white blood cell counts and to study red blood cell morphology. Abnormal granulocyte, lymphocyte or monocyte cell counts may be used to facilitate the diagnosis of diseases such as leukemia or bacterial infections
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.