Learn More
Volu Sol Wright Stain Kit, 8 oz / 250 mL, Volu-Sol™
SDP
Supplier:  Volu Sol VWS300
Encompass_Preferred
Wright and Wright-Giemsa Stain Kits include three components:  a stain, buffer, and a rinse.  Stain peripheral blood and  bone marrow smears. Used to perform differential white blood cell counts and to study red blood cell morphology. Abnormal granulocyte, lymphocyte or monocyte cell counts may be used to facilitate the diagnosis of diseases such as leukemia or bacterial infections
Catalog No. 50-420-780
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.