Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER TITRANT, 5 mg/ml, in METHANOL | 6 X 1 L | GFS CHEMICALS, INC. Item #: 99823 | Bulk #: 1616
SDP
Supplier:  GFS Chemicals Inc 99823
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
WATERMARK KARL FISCHER TITRANT, 5 mg/ml, in METHANOL | Iodine/methanol volumetric titrant. Must be used in conjunction with KF Solvent 1610.; Specifications : Suitability, Titer Strength (mg/ml) 4.90 - 5.10 mg/ml
Catalog No. NC0941438