Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix X Marks the Spot Deluxe Kit Components
SCI_ED

Catalog No. S394031XM Shop All AquaPhoenix Scientific Products
Click to view available options
Product Type:

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Product Type

Mirror

Grade

K to 5

For Use With (Application)

Education kit

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.