Learn More
Cole-Parmer Cole-Parmer X-Ray Film Cassette, Aluminum, 8x10"
Supplier:  Cole-Parmer 2170035
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Get the results you need at an affordable price compared to a name brand blue-sensitive, double-emulsion film for performance and value. These cassetts are ideal for western blotting, DNA sequencing gels, chemiluminescent probes, gel shift analysis, and commonly used radioisotopes (32P, 125I). Its double emulsion film gives you sharp contrast and exemplary resolution for DNA sequencing. The film is single-sided for exceptional clarity and low background.
Catalog No. NC0581965
missing translation for 'QuantityLeft'