Learn More
TARGETMOL CHEMICALS INC XANTHINE AMINE CONGENER 5MG
Supplier:  TARGETMOL CHEMICALS INC T23535L
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Xanthine amine congener trihydrochloride is a potent adenosine antagonist that reverses the reduction in urine flow sodium excretion and heart rate produced by the adenosine agonist N6-cyclohexyladenosine. purity: 97%
Catalog No. 50-258-0863
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.