Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XCC, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Adenosine receptor antagonist (Ki values are 42, 68 and 1130 nM for A1, A2B and A2A receptors).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 386.4 |
Percent Purity | >98% |
Chemical Name or Material | XCC |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction