Learn More
Agilent Technologies XL1-Blue Competent Cells. 5 x 0.2 mL. For routine cloning, ≥ 1 x 108 transformants/µg
Supplier:  Agilent Technologies 200249
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
XL1-Blue Competent Cells, Strain of choice for preparation of high-quality plasmid DNA, Single-strand rescue of phagemid DNA, Contains an antibiotic-resistant F' episome, Available in a wide variety of transformation efficiencies
Catalog No. 50-125-058