Learn More
Agilent Technologies XL1-BLUE SUB GRADE COMP CELLS
Supplier:  Agilent Technologies 200130
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
XL1-Blue Subcloning Grade Competent Cells. For cloning where DNA is not limited, >/=1 x 10^6 transformations/ug. Preparation of high-quality plasmid DNA. Allows blue-white color screening.
Catalog No. 50-125-010
missing translation for 'QuantityLeft'