Learn More
Agilent Technologies XL1-BLUE SUPERCOMPETENT CELLS
Supplier:  Agilent Technologies 200236
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Cell, Supercompetent, XL1-Blue; Stratagene; For high-efficiency cloning, greater than equal1 x 109 transformant/ug; Preparation of quality plasmid DNA; Blue-white color screening; Contain antibiotic-resistant F episome; Versatile cloning
Catalog No. 50-125-053
missing translation for 'QuantityLeft'