Learn More
Agilent Technologies XL10-GOLD KANRULTCOMPCELLS
Supplier:  Agilent Technologies 200317
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
XL10-Gold Kan r Ultracompetent Cells, features kanamycin-resistance gene on the F in. episome, for extremely demanding cloning in chloramphenicol-resistant vectors. Increase the efficiency of introducing eukaryotic DNA into E. coli.
Catalog No. 50-125-096