Learn More
Agilent Technologies XL10-GOLD ULTRACOMP CELLS
Supplier:  Agilent Technologies 200314
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
XL10-Gold ultracompetent cells are created for transformation of large DNA molecules with high efficiency. Ideal for constructing plasmid DNA libraries, as they decrease size bias and produce larger, more complex plasmid libraries.
Catalog No. 50-125-094
missing translation for 'QuantityLeft'