Learn More
Agilent Technologies XL1BLUE MRF E.COMP CELLS
Supplier:  Agilent Technologies 200158
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Cell, Electroporation-Competent, XL1-Blue MRF; Stratagene; For routine electroporation cloning applications; Ideal for cloning methyl; Restriction minus for cloning methylated DNA, greater than equal1 x 1010 transformants/ug
Catalog No. 50-125-021
missing translation for 'QuantityLeft'