Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRF Standard, 0.05% in 70% w/w Phosphoric Acid, Reagents

Catalog No. 01333375
Click to view available options
:

XRF Standard, 0.05% in 70% w/w Phosphoric Acid, Reagent, use for XRF Spectrometry. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material

XRF Standard, 0.05% in 70% w/w Phosphoric Acid

Packaging

Poly Bottle

CAS

7664-38-2, 7732-18-5, 7782-61-8

Quantity

500 mL

Color

Orange

Percent Purity

0.05%

Grade

Reagent

pH

<2

Boiling Point

132°C

Specific Gravity

1.526

Solubility

Completely miscible

Physical Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.