Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRF Standard, 0.40% in 40% w/w Phosphoric Acid, Reagents

Catalog No. 01333376
Click to view available options
:
Quantity: 500 mL

XRF Standard, 0.40% in 40% w/w Phosphoric Acid, Reagent, use for XRF Spectrometry. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material XRF Standard, 0.40% in 40% w/w Phosphoric Acid
Packaging Poly Bottle
CAS 7732-18-5, 7664-38-2, 7782-61-8
Quantity 500 mL
Color Orange
Percent Purity 0.4%
Grade Reagent
pH <2
Boiling Point 113°C
Specific Gravity 1.2778
Solubility Completely miscible
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.