Learn More
Assay Technology 566 Organic Vapor Monitor | 1 chemical | box of 5
SDP
Supplier:  Assay Technology X5661AT
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The 566 Organic Vapor Monitor is our most versatile and popular organic vapor badge. The 566 utilizes our ChemDisk II configuration with a flexible, inert charcoal wafer, making it smaller and much easier to use than other badges on the market. The 76-hole sampling grid allows for sampling rates around 8 mL/min. While the 8-hour reporting limits vary from chemical to chemical (see our Sampling Guide), most are in the 0.1 ppm range, well below most OSHA PELs and NIOSH RELs. In addition, the 566 badge has much more sampling capacity than most other badge and tube methods.
Catalog No. NC9863337
missing translation for 'QuantityLeft'