Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene, Vion Biosciences

Catalog No. p-200076280 Shop All Aldon Products
Click to view available options
Quantity:
WARNING: This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.
WARNING: This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.

This xylene is an excellent choice for a clearing and deparaffinizing agent in your histological processes.

  • This xylene is an excellent choice for a clearing and deparaffinizing agent in your histological processes.

Specifications

Boiling Point

139°C to 141°C (283°F to 286°F)

Color

Colorless

Content And Storage

Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.

Packaging Type

Amber glass bottle

Melting Point

Data not available.

Product Line

Vion Histology

Product Type

Clearing Agent

Quantity

500 mL

Formulation

100% Xylenes

For Use With (Application)

Histology

Reagent Type

Solvent

Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.