Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YE 120, Tocris Bioscience™

Catalog No. p-4816904 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GPR35 agonist

GPR35 agonist (EC50 = 32 nM in DMR assays). Also displays partial agonist activity in a β-arrestin translocation assay (EC50 = 10.2 μM).

Chemical Identifiers

CAS 383124-82-1
Molecular Formula C16H9Cl2N3O
Molecular Weight (g/mol) 330.168
InChI Key RSWZFAPYKOARNG-UHFFFAOYSA-N
Synonym 2-3-cyano-5-3,4-dichlorophenyl-4,5-dimethyl-2 5h-furanylidene propanedinitrile, 2-3-cyano-5-3,4-dichlorophenyl-4,5-dimethylfuran-2-ylidene propanedinitrile
PubChem CID 12138435
IUPAC Name 2-[3-cyano-5-(3,4-dichlorophenyl)-4,5-dimethylfuran-2-ylidene]propanedinitrile
SMILES CC1=C(C(=C(C#N)C#N)OC1(C)C2=CC(=C(C=C2)Cl)Cl)C#N

Specifications

Quantity 10 mg
Formula Weight 330.17
Percent Purity >98%
Chemical Name or Material YE 120
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.