Learn More
Clontech Labs 3P YEAST MEDIA-URA DOFU 10GR
SDP
Supplier:  Clontech Labs 3P 630416
Encompass
Ura DO Supplement 10 g various mixtures of amino acids and nucleosides to add to the Minimal SD Base to make a defined minimal medium for growing yeast cultures confirming auxotrophic phenotypes and selecting yeast transformants
Catalog No. NC9413251
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.