Learn More
Clontech Labs 3P Minimal SD Base, 267 g
SDP
Supplier:  Clontech Labs 3P 630411
Encompass
These high-quality yeast growth media and dropout supplements can be used for growing yeast cultures, confirming auxotrophic phenotypes, and selecting yeast transformants. 267 g
Catalog No. NC9419268
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.