Learn More
Zymo Research Corporation Zymoprep Yeast Plasmid Miniprep II, 50 Preps
Supplier: Zymo Research Corporation D2004
The Zymoprep™ Yeast Plasmid Miniprep II provides all the necessary reagents for plasmid isolation from S. cerevisiae, C. albicans and S. pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The procedure is simple and efficient, and there is no need for glass beads or phenol. Reliably recover plasmid DNA from yeast colonies, patches on plates, or as liquid cultures. The system is ideal for low-copy number and hard to isolate plasmids. Eluted plasmid DNA can be used directly for E. Coli transformation, PCR, and Southern blot analysis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.