Learn More
Bioworld YT broth (2x), 100 g
Supplier: Bioworld 306201031
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.Contains (g/L): Tryptone: 16 g Yeast Extract 10g Sodium Chloride 5gInstruction to Use: Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final ph@ 25°C: 6.8-7.2. This product is sold for laboratory research use only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.