Learn More
Bioworld YT broth (2x), 5 kg
Supplier:  Bioworld 306201034
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.Contains (g/L): Tryptone: 16 g Yeast Extract 10g Sodium Chloride 5gInstruction to Use: Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final ph@ 25°C: 6.8-7.2. This product is sold for laboratory research use only.
Catalog No. 50-254-405
missing translation for 'QuantityLeft'