Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-7051543
Click to view available options
Quantity:
25 g
100 g

Rare earth element doped yttrium aluminum oxide is used in solid state lasers

Rare earth element doped yttrium aluminum oxide is used in solid state lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12005-21-9
MDL Number MFCD00213958
Synonym Yttrium aluminum garnet; YAG

Specifications

Melting Point 1760°C
Physical Form Powder
Quantity 25 g
Formula Weight 593.62
Percent Purity 99.9%
Refractive Index 1.833
Density 4.65 g/mL
Assay (REO)
Chemical Name or Material Yttrium aluminum oxide
EINECSNumber 234-465-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.