Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog No. p-4408036
Click to view available options
Quantity:
25 g
100 g

CAS: 85949-60-6 | C6H17O10Y | 338.10 g/mol

Yttrium(III) acetate tetrahydrate is used as laboratory reagents, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85949-60-6
Molecular Formula C6H17O10Y
Molecular Weight (g/mol) 338.10
MDL Number MFCD00013048
InChI Key AIQRTHPXPDTMBQ-UHFFFAOYSA-K
IUPAC Name yttrium(3+) triacetate tetrahydrate
SMILES O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in Hot water.
Formula Weight 338.10 (266.04 Anhydrous)
Density 1.5 g/mL
Chemical Name or Material Yttrium(III) acetate tetrahydrate
EINECSNumber 245-612-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.