Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-7050162
Click to view available options
Quantity:
1 g
5 g

CAS: 13470-38-7

Yttrium(III) iodide, is used as a starting material for YBCO superconducting materials in a lower temperature preparation. Applied in the solid-state synthesis of a new compound, Y21I18C14B7, which is the first example of a rare earth metal halide with interstitial halogen atoms with misfit characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 13470-38-7
MDL Number MFCD00054144

Specifications

Melting Point 1,004°C
Physical Form Chunks and/or Powder
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 469.62
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Yttrium(III) iodide, Ultra dry
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-737-1
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.