Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) nitrate hexahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-4412870
Click to view available options
Quantity:
5 g
25 g
100 g
5 x 100 g

CAS: 13494-98-9 | H12N3O15Y | 383.008 g/mol

It is used as a reagent, in optics, ceramics, glass and electronics. It is used in superconducting materials. It has been reported as a reusable catalyst in Bignelli reaction for the synthesis of heterocycles such as pyrimidin-2-ones. It is a powerful catalyst for the synthesis of some supramolecules such as calix[4]resorcinarenes, 1,8-dioxooctahydroxanthenes, 2-amino-4H-chromenes, and several other organic condensation products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13494-98-9
Molecular Formula H12N3O15Y
Molecular Weight (g/mol) 383.008
MDL Number MFCD00149940
InChI Key QBAZWXKSCUESGU-UHFFFAOYSA-N
Synonym yttrium nitrate hexahydrate, yttrium iii nitrate hexahydrate, yttriumnitrat german, yttrium iii nitrate hexahydrate 1:3:6, yttrium trinitrate hexahydrate, nitric acid, yttrium 3+ salt, hexahydrate, nitric acid,yttrium 3+ salt, hexahydrate 8ci,9ci, yttriumnitrat, yttrium iii nitrate hexahydrate reo, ytterium nitrate
PubChem CID 166833
IUPAC Name yttrium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Y+3]

Specifications

Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 383.01 (274.92 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Yttrium(III) nitrate hexahydrate
Hazard Category H302-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 233-802-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.