Learn More
Strem, An Ascensus Company CAS# 7440-65-5. 5g. Yttrium powder (99.9% REO). MFCD00011468
SDP
Supplier:  Strem, An Ascensus Company 9339405G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CAS# 7440-65-5. 5g. Yttrium powder (99.9% REO). MFCD00011468. Molecular Weight: 88.90. Molecular Formula: Y. Color/form: -40 mesh Strem# 93-3940. http://www.strem.com/catalog/v/93-3940/
Catalog No. 50-901-15232