Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HORIBA Yumizen C1200 Urine Control

Catalog No. p-7213523 Shop All ABX Diagnostics, Inc. Products
This item is not returnable. View return policy
This item is not returnable. View return policy

Yumizen C1200 Urine Control used to monitor urine assays

  • Used to monitor urine assays

Specifications

Certifications/Compliance

CE Marked

Content And Storage

2°C to 8°C

Concentration

Normal concentration for analytes

Detectable Analytes

Amylase, Calcium, Creatinine, Glucose, Magnesium, Phosphate, Urea, UA, Urinary Proteins

For Use With (Equipment)

Used on Yumizen C1200 Clinical Chemistry Analyzer

Sample Type

Urine

Type

Control

Disposable

Single-use

Quantity

6 Pk. 5 mL Vials

Source

Human Urine, Human and Bovine Serum Albumin

Stability

Stable for 3 months (at 2°C to 8°C)

Sterility

Non-sterile

Form

Liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.