Learn More
Medchemexpress LLC HY-106004 10mg Medchemexpress, Zamicastat CAS:1080028-80-3 Purity:>98%
Supplier: Medchemexpress LLC HY10600410MG
Medchemexpress, HY-106004 10mg Zamicastat CAS:1080028-80-3 Zamicastat (BIA 5-1058) is a dopamine β-hydroxylase (DBH) inhibitor that could cross the blood-brain barrier (BBB) and cause central as well as peripheral effects. Zamicastat is also a concentration-dependent dual P-gp and BCRP inhibitor with IC50 values of 73.8 μM and 17.0 μM, respectively. Reduces high blood pressure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.